Learn More
Occludin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13409-1-AP-150UL
 View more versions of this product
Product Code. 16891604
  • £349.00 / 150µL
Description
This gene encodes an integral membrane protein that is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Mutations in this gene are thought to be a cause of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG), an autosomal recessive neurologic disorder that is also known as pseudo-TORCH syndrome. Alternative splicing results in multiple transcript variants. A related pseudogene is present 1.5 Mb downstream on the q arm of chromosome 5.
Specifications
Specifications
Occludin
Polyclonal
Unconjugated
OCLN
occludin, OCLN
Rabbit
Antigen Affinity Chromatography
RUO
100506658, 18260, 83497
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16625, Q61146, Q6P6T5
Ocln
Occludin Fusion Protein Ag4057
150 μL
Primary
Human, Mouse, Rat