missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCAM/NCAM2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
£464.80
Specifications
Antigen | OCAM/NCAM2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
OCAM/NCAM2 Polyclonal specifically detects OCAM/NCAM2 in Mouse samples. It is validated for Western Blot.Specifications
OCAM/NCAM2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
NCAM2, N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10, Rb-8, RNCAM | |
NCAM2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O35136 | |
4685 | |
Mouse myeloma cell line NS0-derived recombinant mouse OCAM Leu20-Gly700 Accession # O35136 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse OCAM in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) NCAM-1 is observed. |
For Research Use Only