missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCAM/NCAM2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF778
This item is not returnable.
View return policy
Description
OCAM/NCAM2 Polyclonal specifically detects OCAM/NCAM2 in Mouse samples. It is validated for Western Blot.
Specifications
| OCAM/NCAM2 | |
| Polyclonal | |
| Unconjugated | |
| O35136 | |
| NCAM2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OCAM Leu20-Gly700 Accession # O35136 | |
| 100 μg | |
| Primary | |
| Detects mouse OCAM in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) NCAM-1 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NCAM2, N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10, Rb-8, RNCAM | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4685 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction