Learn More
Agilent BioTek Objectives for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
Brand: Agilent BioTek 1220549
Additional Details : Weight : 0.50000kg
Description
Select from a range of objectives that provide 2.5x to 60x imaging.Specifications
Objective 2.5X (2.25x eff.), NA: .07 | |
Cytation3 Cell Imaging Multi-Mode Reader | |
For use only in the blue and green emission ranges. Not compatible with red emission filter cubes. |