Learn More
Agilent BioTek Objectives for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
Brand:  Agilent BioTek 1220544
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15458712
  • £7390.25 / Each
Estimated Shipment: 28-05-2024
to see stock.
Description
Description
Select from a range of objectives that provide 2.5x to 60x imaging.
Specifications
Specifications
Objective 40X, NA: .6
Imaging Multi-Mode Reader