missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OATP1B1/OATP2 Antibody (ESL), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£169.00
Specifications
| Antigen | OATP1B1/OATP2 |
|---|---|
| Clone | ESL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunofluorescence 1:100 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
OATP1B1/OATP2 Monoclonal specifically detects OATP1B1/OATP2 in Human, Canine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
| OATP1B1/OATP2 | |
| Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunofluorescence 1:100 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human | |
| Liver-specific organic anion transporter 1, LST1SLC21A6, LST-1Solute carrier family 21 member 6, OATP1B1Sodium-independent organic anion-transporting polypeptide 2, OATP2MGC133282, OATP-COATP-2, OATPCsolute carrier organic anion transporter family member 1B1, solute carrier family 21 (organic anion transporter), member 6, solute carrier organic anion transporter family, member 1B1 | |
| SLCO1B1 | |
| IgM | |
| Unpurified | |
| OATP2 (ESL) |
| ESL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| Tissue culture supernatant with 0.09% Sodium Azide | |
| 10599 | |
| Synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title