missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OATP1B1/OATP2 Antibody (ESL), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-74481-0.025ml
This item is not returnable.
View return policy
Description
OATP1B1/OATP2 Monoclonal specifically detects OATP1B1/OATP2 in Human, Canine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications
| OATP1B1/OATP2 | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with 0.09% Sodium Azide | |
| SLCO1B1 | |
| Synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2. | |
| 0.025 mL | |
| Membrane Trafficking and Chaperones | |
| 10599 | |
| Human | |
| Supernatant |
| Western Blot | |
| ESL | |
| Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunofluorescence 1:100 | |
| Liver-specific organic anion transporter 1, LST1SLC21A6, LST-1Solute carrier family 21 member 6, OATP1B1Sodium-independent organic anion-transporting polypeptide 2, OATP2MGC133282, OATP-COATP-2, OATPCsolute carrier organic anion transporter family member 1B1, solute carrier family 21 (organic anion transporter), member 6, solute carrier organic anion transporter family, member 1B1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| OATP2 (ESL) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction