missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-D-serine methyl ester hydrochloride, 98%, Thermo Scientific™

Product Code. 15458566
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458566 25 g 25g
15448566 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458566 Supplier Thermo Scientific Alfa Aesar Supplier No. H63874.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78537-14-1
Molecular Formula C8H18ClNO3
Molecular Weight (g/mol) 211.686
MDL Number MFCD00077108
InChI Key PCIABNBULSRKSU-FYZOBXCZSA-N
Synonym h-d-ser tbu-ome.hcl, h-d-ser tbu-ome hydrochloride, o-t-butyl-d-serine methyl ester hydrochloride, ser tbu-ome.hcl, r-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, h-d-ser tbu-ome hcl, o-tert-butyl-d-serine methyl ester hydrochloride, methyl o-tert-butyl-d-serinate-hydrogen chloride 1/1, methyl 2r-2-amino-3-tert-butoxy propanoate hydrochloride
PubChem CID 13571432
IUPAC Name methyl (2R)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride
SMILES CC(C)(C)OCC(C(=O)OC)N.Cl

Specifications

Color White
Beilstein 2041979
Quantity 25 g
Formula Weight 211.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material O-tert-Butyl-D-serine methyl ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.