CAS RN 78537-14-1
IUPAC Name:
methyl 2-amino-3-(tert-butoxy)propanoate hydrochloride
Synonyms:
methyl 2-amino-3-(tert-butoxy)propanoate hydrochloride
Molecular Weight (g/mol):
211.69
Molecular Formula:
C8H18ClNO3
InChi Key:
PCIABNBULSRKSU-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)C(N)COC(C)(C)C
1
–
1
of
1
results
1
O-tert-Butyl-D-serine methyl ester hydrochloride, 98%, Thermo Scientific™
CAS: 78537-14-1 Molecular Formula: C8H18ClNO3 Molecular Weight (g/mol): 211.686 MDL Number: MFCD00077108 InChI Key: PCIABNBULSRKSU-FYZOBXCZSA-N Synonym: h-d-ser tbu-ome.hcl,h-d-ser tbu-ome hydrochloride,o-t-butyl-d-serine methyl ester hydrochloride,ser tbu-ome.hcl,r-methyl 2-amino-3-tert-butoxy propanoate hydrochloride,h-d-ser tbu-ome hcl,o-tert-butyl-d-serine methyl ester hydrochloride,methyl o-tert-butyl-d-serinate-hydrogen chloride 1/1,methyl 2r-2-amino-3-tert-butoxy propanoate hydrochloride PubChem CID: 13571432 IUPAC Name: methyl (2R)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride SMILES: CC(C)(C)OCC(C(=O)OC)N.Cl