Learn More
DWK Life Sciences O-Ring, suitable only for desiccator Base with ring-grooved flange, from silicone (VMQ)
Product Code. 10663982 Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
For Models From 1996
Unit Size:
Each
Product Code. 10663982
Brand: DWK Life Sciences 292156108
Only null left
Add to basket
Add to basket
The O-ring is a seal suitable for the desiccator with fluted bevel and enables a tight seal.
The O-ring is a seal suitable for the desiccator with fluted bevel, which seals the desiccator tightly with a precisely fitting shape. The use of the seal eliminates the need for grinding grease.
  • With the help of the O-ring, a tight seal of the desiccator with annular groove flange can be achieved
  • Suitable only for desiccator base with ring-grooved flange
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Tight closure of desiccators; Ordering advice: the O-ring is dimensioned according to the nominal doameter (DN)
For Use With (Equipment) For Models From 1996
Quantity 1 Ea.
Material Silicone
Diameter (Metric) Outside 236 mm