missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NYX Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £366.00
Specifications
| Antigen | NYX |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665632
|
Novus Biologicals
NBP2-94226-0.02ml |
0.02 mL |
£150.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638372
|
Novus Biologicals
NBP2-94226-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NYX Polyclonal antibody specifically detects NYX in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| NYX | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 60506 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CLRP;CSNB1;CSNB1A;CSNB4;NBM1;Nyctalopin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 282-481 of human NYX (NP_072089.1). LLYLDRNSIAFVEEGAFQNLSGLLALHLNGNRLTVLAWVAFQPGFFLGRLFLFRNPWCCDCRLEWLRDWMEGSGRVTDVPCASPGSVAGLDLSQVTFGRSSDGLCVDPEELNLTTSSPGPSPEPAATTVSRFSSLLSKLLAPRVPVEEAANTTGGLANASLSDSLSSRGVGGAGRQPWFLLASCLLPSVAQHVVFGLQMD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title