missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NYX Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94226-0.02ml
This item is not returnable.
View return policy
Description
NYX Polyclonal antibody specifically detects NYX in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NYX | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| CLRP;CSNB1;CSNB1A;CSNB4;NBM1;Nyctalopin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 282-481 of human NYX (NP_072089.1). LLYLDRNSIAFVEEGAFQNLSGLLALHLNGNRLTVLAWVAFQPGFFLGRLFLFRNPWCCDCRLEWLRDWMEGSGRVTDVPCASPGSVAGLDLSQVTFGRSSDGLCVDPEELNLTTSSPGPSPEPAATTVSRFSSLLSKLLAPRVPVEEAANTTGGLANASLSDSLSSRGVGGAGRQPWFLLASCLLPSVAQHVVFGLQMD | |
| 0.02 mL | |
| Neuroscience | |
| 60506 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction