missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Nutrient Dehydrated Media Pad Sets
Brand: Sartorius™ 14091--47------N
209.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Schaufus Pottinger | |
Dehydrated Culture Media | |
Green | |
47 mm |