missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUPR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NUPR1 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16804044
|
Proteintech
15056-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16894034
|
Proteintech
15056-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The human p8 gene encodes a nuclear protein with strong mitogenic activity, which is expressed in the pancreas in response to acute pancreatitis. Response to TNF alpha and staurosporine-induced apoptosis is not modified by p8 expression. However, cells expressing p8 grow significantly and more rapidly.Specifications
| NUPR1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O60356 | |
| 26471 | |
| NUPR1 Fusion Protein Ag7039 | |
| Primary | |
| -20°C | |
| NUPR1 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Candidate of metastasis 1, COM1, nuclear protein 1, NUPR1, P8, Protein p8 | |
| NUPR1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title