missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUPR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15056-1-AP-150UL
This item is not returnable.
View return policy
Description
The human p8 gene encodes a nuclear protein with strong mitogenic activity, which is expressed in the pancreas in response to acute pancreatitis. Response to TNF alpha and staurosporine-induced apoptosis is not modified by p8 expression. However, cells expressing p8 grow significantly and more rapidly.Specifications
| NUPR1 | |
| Polyclonal | |
| Unconjugated | |
| NUPR1 | |
| Candidate of metastasis 1, COM1, nuclear protein 1, NUPR1, P8, Protein p8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26471 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60356 | |
| NUPR1 | |
| NUPR1 Fusion Protein Ag7039 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction