missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUPL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | NUPL1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NUPL1 Polyclonal antibody specifically detects NUPL1 in Rat samples. It is validated for Western BlotSpecifications
| NUPL1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9818 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| KIAA0410PRO2463, nucleoporin like 1, nucleoporin p58/p45, Nucleoporin-like protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human NUPL1 (NP_054808.1). AAQLQSIHENVKVLKEQYLGYRKMFLGDAVDVFETRRAEAKKWQNTPRVTTGPTPFSTMPNAAAVAMAATLTQQQQPATGPQPSLGVSFGTPFGSGIGTGL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title