missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUPL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94283-0.1ml
This item is not returnable.
View return policy
Description
NUPL1 Polyclonal antibody specifically detects NUPL1 in Rat samples. It is validated for Western Blot
Specifications
| NUPL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA0410PRO2463, nucleoporin like 1, nucleoporin p58/p45, Nucleoporin-like protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human NUPL1 (NP_054808.1). AAQLQSIHENVKVLKEQYLGYRKMFLGDAVDVFETRRAEAKKWQNTPRVTTGPTPFSTMPNAAAVAMAATLTQQQQPATGPQPSLGVSFGTPFGSGIGTGL | |
| 0.1 mL | |
| Signal Transduction | |
| 9818 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction