missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP160 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | NUP160 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610722
|
Novus Biologicals
NBP2-94281-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686642
|
Novus Biologicals
NBP2-94281-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUP160 Polyclonal antibody specifically detects NUP160 in Human samples. It is validated for Western BlotSpecifications
| NUP160 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 23279 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp686M14102, FLJ22583, KIAA0197MGC150678, MGC150679,160 kDa nucleoporin, nuclear pore complex protein Nup160, nucleoporin 160kD, nucleoporin 160kDa, Nucleoporin Nup160, NUP120 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-150 of human NUP160 (NP_056046.1). ALERSFVELSGAERERPRHFREFTVCSIGTANAVAGAVKYSESAGGFYYVESGKLFSVTRNRFIHWKTSGDTLELMEESLDINLLNNAIRLKFQNCSVLPGGVYVSETQNR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title