missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP160 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94281-0.02ml
This item is not returnable.
View return policy
Description
NUP160 Polyclonal antibody specifically detects NUP160 in Human samples. It is validated for Western Blot
Specifications
| NUP160 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686M14102, FLJ22583, KIAA0197MGC150678, MGC150679,160 kDa nucleoporin, nuclear pore complex protein Nup160, nucleoporin 160kD, nucleoporin 160kDa, Nucleoporin Nup160, NUP120 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-150 of human NUP160 (NP_056046.1). ALERSFVELSGAERERPRHFREFTVCSIGTANAVAGAVKYSESAGGFYYVESGKLFSVTRNRFIHWKTSGDTLELMEESLDINLLNNAIRLKFQNCSVLPGGVYVSETQNR | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 23279 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction