Learn More
Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps
Allow constant airflow and minimize chance of contamination by using individually packed Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Brand: Thermo Scientific™ 146003
Code : 76
Additional Details : Weight : 0.80500kg
Description
Allow constant airflow and minimize chance of contamination with Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Nunc sterile filter caps are made with HDPE and Thermo Scientific™ GORE™ dPTFE membrane.
- Filter membrane provides effective barrier against microorganisms
- Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
- Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
- Individually wrapped to ensure sterility
Specifications
White | |
Barcoded Flasks 178983 and 132920 | |
100 | |
Sterile | |
Filter |
HDPE | |
100 | |
Fewer resources used and less waste, Sustainable packaging | |
Class VI |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.