Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
Description
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
- HDPE connector
- Two cover caps
- Two blue sealing caps
- No. 4210 bacterial air vent filter
- Tubing and clamp
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
| Description | Cell Factory System Start-up Kit |
| For Use With (Equipment) | Cell Factory Systems (All) |
| Includes |
|
| No. per Case | 1 |
| No. per Pack | 1 |
| Product Type | System Start-Up Kit |
| Bottom Shape | Flat |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.