missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 1536-Well Microplates Plates
Screen four times as many samples in a single plate with Thermo Scientifi™ Nunc™ 1536-Well Microplates, which are ideal for high-throughput screening.
£1333.57 - £6610.00
Specifications
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
---|---|
Format | Standard |
Lid | Without Lid |
Array | 32 x 48 |
Product Line | Nunc |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11730483
|
Thermo Scientific™
253601 |
Black |
£6610.00
Pack of 90 |
In Stock
Log in to see stock. |
|||||
11321585
|
Thermo Scientific™
253607 |
White |
£5740.00
Pack of 90 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
11331585
|
Thermo Scientific™
253614 |
Clear |
£4885.00
Pack of 90 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
10654502
|
Thermo Scientific™
253607 |
White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10409701
|
Thermo Scientific™
253614 |
Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Maximize high-throughput screening efficiency by using Thermo Scientific™ Nunc™ 1536-Well Microplates, built according to ANSI standards and with the highest quality.
- Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum signal reflection and minimal crosstalk between wells
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
Nunc | |
Polystyrene | |
1 to 10 μL | |
Non-treated | |
90 | |
90/Cs. |
Standard | |
32 x 48 | |
Microplate | |
1536 | |
Non-sterile | |
Flat Bottom (F-well) | |
30 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.