missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Numb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£156.00 - £301.00
Specifications
| Antigen | Numb |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-2ug/ml, Immunohistochemistry reported in scientific literature (PMID 30256757), Immunohistochemistry-Paraffin 5-10ug/ml∼ |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18193245
|
Novus Biologicals
NBP2-27181SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108372
|
Novus Biologicals
NBP2-27181 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Numb Polyclonal specifically detects Numb in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Numb | |
| Western Blot 1-2ug/ml, Immunohistochemistry reported in scientific literature (PMID 30256757), Immunohistochemistry-Paraffin 5-10ug/ml∼ | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| NUMB | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen for the NUMB antibody is 100% homologous in human, mouse, rat, cow and monkey. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine, Primate | |
| P49757 | |
| 8650 | |
| A synthetic peptide corresponding to amino acids 12-40 of human NUMB was used as the immunogen for the antibody, GenBank no gb AAH20788.1 . | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title