missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Numb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27181SS
This item is not returnable.
View return policy
Description
Numb Polyclonal specifically detects Numb in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Numb | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the NUMB antibody is 100% homologous in human, mouse, rat, cow and monkey. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1-2ug/ml, Immunohistochemistry reported in scientific literature (PMID 30256757), Immunohistochemistry-Paraffin 5-10ug/ml∼ | |
| P49757 | |
| NUMB | |
| A synthetic peptide corresponding to amino acids 12-40 of human NUMB was used as the immunogen for the antibody, GenBank no gb AAH20788.1 . | |
| 0.025 mg | |
| Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 8650 | |
| Human, Mouse, Rat, Bovine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction