missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54929
This item is not returnable.
View return policy
Description
NUDT13 Polyclonal specifically detects NUDT13 in Human samples. It is validated for Western Blot.
Specifications
| NUDT13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZp586P2219, EC 3.-, nucleoside diphosphate-linked moiety X motif 13, nudix (nucleoside diphosphate linked moiety X)-type motif 13, Nudix motif 13, Protein KiSS-16 | |
| Rabbit | |
| 40 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q86X67 | |
| NUDT13 | |
| Synthetic peptides corresponding to NUDT13(nudix (nucleoside diphosphate linked moiety X)-type motif 13) The peptide sequence was selected from the N terminal of NUDT13. Peptide sequence MSLYCGIACRRKFFWCYRLLSTYVTKTRYLFELKEDDDACKKAQQTGAFY. | |
| Affinity purified | |
| RUO | |
| 25961 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction