missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Rabbit anti-Human, Mouse, Rat, Clone: 9C8T10, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Nudel |
|---|---|
| Clone | 9C8T10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325802
|
Novus Biologicals
NBP3-16275-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18312866
|
Novus Biologicals
NBP3-16275-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nudel Monoclonal antibody specifically detects Nudel in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Nudel | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 81565 | |
| IgG | |
| Affinity purified |
| 9C8T10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Nudel (Q9GZM8). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title