missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Rabbit anti-Human, Mouse, Rat, Clone: 9C8T10, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16275-100UL
This item is not returnable.
View return policy
Description
Nudel Monoclonal antibody specifically detects Nudel in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Nudel | |
| Monoclonal | |
| Unconjugated | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Nudel (Q9GZM8). | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 81565 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 9C8T10 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction