missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress NucPE1
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Specifications
Specifications
| Content And Storage | 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen) |
| Product Type | Dye Reagents |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 25 mg/mL (51.72 mM; Need ultrasonic) |
| Molecular Weight (g/mol) | 483.36 g/mol |
| CAS | 1404091-23-1 |
| Synonym | Nuclear Peroxy Emerald 1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction