missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress NucPE1

Product Code. 16490007
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490007

Brand: MedChemExpress HY101859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NucPE1 (Nuclear Peroxy Emerald 1) is a nuclear-localized fluorescent hydrogen peroxide that is specifically localized to cellular nuclei without appended targeting moieties.

Specifications

Content And Storage 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Product Type Dye Reagents
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (51.72 mM; Need ultrasonic)
Molecular Weight (g/mol) 483.36 g/mol
CAS 1404091-23-1
Synonym Nuclear Peroxy Emerald 1
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.