missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ TriPrep, Mini kit for RNA, DNA, and Protein Purification

Product Code. 11962502
Change view
Click to view available options
Sufficient For:
10rxns
250rxns
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
11962502 10rxns 10 reactions
11972392 - 50 reactions
11962262 250rxns 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11962502 Supplier Macherey-Nagel™ Supplier No. 740966.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Parallel isolation of RNA, DNA and, protein from an undivided sample.

NucleoSpin™ miRNA is suitable for the parallel isolation of small RNA, large RNA and protein.

Superior selectivity - purify your RNA fractionated by size

Isolation of small RNA only (18 to 200 bases)Isolation of small (18 to 200 bases) and large (>200 bases) RNA in two fractionsIsolation of total RNA (small and large in one fraction)

Additional isolation of total protein

Denatured protein ready-to-use for analysis in SDS-PAGE and Western blot.

Excellent RNA recovery - even without phenol/chloroform

Lysis with chaotropic saltSpin column based procedure

Convenient handling - complete equipment provided

NucleoSpin filters - efficient sample homogenisationNucleoSpin protein removal columns - highest purity of small RNArDNase - highly efficient on-column digestion of genomic DNA at RT

  • Silica membrane technology
  • Format: minispin columns
  • Sample size: <107 cultured cells, <30mg human/tissue; <50 mg plant tissue, <150μL reaction mixture
  • Fragment size, small RNA: 18 to 200 bases
  • Fragment size, large RNA: >200 bases
  • Typical yield: 10μg small RNA; 95μg large RNA from 107 HeLa cells
  • Binding capacity: 200μg
  • Elution volume, 30μL to 100μL
  • Preparation time: <45min (6 preps human/animal tissue, small and large RNA); <35min (6 preps human/animal tissue, small RNA only)
  • Small RNA (18 to 200 bases)
  • Large RNA (>200 bases)
  • Total RNA (small and large RNA in one fraction), and isolate your total protein fraction For small and large RNA and total RNA purification, and isolation of total protein fraction.

Specifications

Product Type NucleoSpin™ TriPrep Column Set
Quantity 1 Set
Includes 2mL Collection Tubes, 1.5mL Collection Tubes, NucleoSpin™ Filters, buffers, RNase-free and rDNase
Sufficient For 10rxns
Column Type Spin Column
For Use With (Application) For Rapid Purification of Total RNA, DNA, and Protein From Small and Precious Samples
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.