missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles

Product Code. 10311625 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Flow Rate:
0.15 mL/min
0.34 mL/min
0.6 mL/min
0.8 mL/min
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Flow Rate unitSize
10311625 125 mm 2 mm 0.15 mL/min Each
10668614 100 mm 2 mm 0.15 mL/min Each
10711325 250 mm 2 mm 0.15 mL/min Each
12738862 50 mm 2 mm 0.15 mL/min Each
10480864 125 mm 3 mm 0.34 mL/min Each
12728862 250 mm 3 mm 0.34 mL/min Each
12717850 100 mm 4.6 mm 0.8 mL/min Each
12748862 50 mm 3 mm 0.34 mL/min Each
12408467 125 mm 4.6 mm 0.8 mL/min Each
12428467 150 mm 4.6 mm 0.8 mL/min Each
12488457 250 mm 4.6 mm 0.8 mL/min Each
12707850 50 mm 4.6 mm 0.8 mL/min Each
10627715 250 mm 4 mm 0.6 mL/min Each
12498457 125 mm 4 mm 0.6 mL/min Each
12418467 50 mm 4 mm 0.6 mL/min Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 10311625 Supplier Macherey-Nagel™ Supplier No. 760531.20
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for analytical HPLC applications with hydrophilic compounds. Macherey-Nagel™ NUCLEODUR™ HILIC Column for HPLC, 3μm Particles is an EC standard column with high purity silica phase. The columns are available in a variety of diameters and lengths.

This product is suitable for Hydrophilic Interaction Chromatography (HILIC) applications. It features a highly pure ammonium/sulfonic acid-modified silica phase with an 80:20 acetonitrile-water eluent. These EC standard columns have a particle size of 3μm and are used in analytical HPLC.

  • Ideal for reproducible and stable chromatography of highly polar analytes
  • Suitable for analytical and preparative applications
  • Very short column conditioning period
  • Used with hydrophilic compounds, such as: organic polar acids and bases, polar natural compounds, nucleosides, oligonucleotides, amino acids, peptides, water soluble vitamins

Specifications

Column Chemistry Ammonium /sulfonic acid
Column Format Analytical Column
Crosslinking Ammonium /sulfonic acid
Diameter (Metric) 2 mm
Flow Rate 0.15 mL/min
For Use With For Reproducible and Stable Chromatography of Highly Polar Analytes
Functionality/Interaction Type Hydrophobic
Hydrophobicity Yes
Length (Metric) 125 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility 80:20 Acetonitrile/water
Particle Shape Spherical
Particle Size 3 μm
pH 2 to 8.5
Pore Size 110 Å
Pore Volume 0.9 mL/g
Quantity 1 Ea.
Solvents 80:20 Acetonitrile/water
Surface Area 340 m²/g
Temperature 60°C
Stationary Phase Ammonium/Sulfonic Acid Modified Silica
Packing Material Silica
Carbon Load 7%
Column Type HILIC
Endcapped No
Phase HILIC
Material Stainless Steel
Type EC Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.