missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Pore Complex Protein Nsp1p Antibody (32D6), Novus Biologicals™
Mouse Monoclonal Antibody
£153.00 - £342.00
Specifications
| Antigen | Nuclear Pore Complex Protein Nsp1p |
|---|---|
| Clone | 32D6 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Supernatant |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18291145
|
Novus Biologicals
NBP1-05227-0.125ML |
0.125 mL |
£153.00
125µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253520
|
Novus Biologicals
NBP1-05227 |
0.5 mL |
£342.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuclear Pore Complex Protein Nsp1p Monoclonal specifically detects Nuclear Pore Complex Protein Nsp1p in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Nuclear Pore Complex Protein Nsp1p | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| 107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP153, NUP155, NUP84, NUP98 | |
| IgG1 | |
| Tissue culture supernatant | |
| 86.5 kDa |
| 32D6 | |
| Unconjugated | |
| Mouse | |
| Yeast | |
| Yeast nuclear preparations | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title