missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Pore Complex Protein Nsp1p Antibody (32D6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05227
This item is not returnable.
View return policy
Description
Nuclear Pore Complex Protein Nsp1p Monoclonal specifically detects Nuclear Pore Complex Protein Nsp1p in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Nuclear Pore Complex Protein Nsp1p | |
| Monoclonal | |
| Unconjugated | |
| Supplied as concentrated hybridoma cell culture media. with 5mM Sodium Azide | |
| Mouse | |
| 86.5 kDa | |
| 0.5 mL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 32D6 | |
| Western Blot 1:10000-1:50000, Immunohistochemistry 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000, Immunoprecipitation | |
| 107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP153, NUP155, NUP84, NUP98 | |
| Yeast nuclear preparations | |
| Tissue culture supernatant | |
| RUO | |
| Yeast | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction