missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Rabbit anti-Human, Primate, Clone: 235-1R, Novus Biologicals™
Rabbit Monoclonal Antibody
£238.00 - £439.00
Specifications
| Antigen | Nuclear Antigen |
|---|---|
| Clone | 235-1R |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen 1-2 ug/ml |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018235
|
Novus Biologicals
NBP3-13912-20UG |
20 μg |
£238.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18316602
|
Bio-Techne
NBP3-13912-100UG |
100 μg |
£439.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
| Nuclear Antigen | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Human, Primate | |
| Nuclei, Nucleus | |
| IgG κ | |
| Protein A or G purified |
| 235-1R | |
| Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Nuclei of human myeloid leukemia biopsy cells | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title