missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Antigen Rabbit anti-Human, Primate, Clone: 235-1R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-13912-100UG
This item is not returnable.
View return policy
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
| Nuclear Antigen | |
| Monoclonal | |
| 0.2 mg/ml | |
| Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen 1-2 ug/ml | |
| Nuclei, Nucleus | |
| Nuclei of human myeloid leukemia biopsy cells | |
| 100 μg | |
| Primary | |
| Store at 4°C. | |
| IgG κ |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| 235-1R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction