Learn More
Invitrogen™ NTRK2 (TRKB) Recombinant Human Protein
NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system.
Brand: Invitrogen™ PR5995D
Code : NEW
Additional Details : Weight : 0.01000kg
Description
NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses.
Specifications
NP_006171 | |
50mM Tris-HCl, pH 7.5, 300mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
NTRK2 (TRKB) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
40.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
Neurotrophic Receptor (NTRK) Kinase Family | |
Recombinant | |
Baculovirus |