missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTRK2 (TRKB) Recombinant Human Protein
NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system.
£1602.00 - £6900.00
Specifications
Accession Number | NP_006171 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 300mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 40.9kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448463
|
Invitrogen™
PR5995D |
100μg |
£1602.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438463
|
Invitrogen™
PR5995A |
1mg |
£6900.00
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses.
Specifications
NP_006171 | |
50mM Tris-HCl, pH 7.5, 300mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
Neurotrophic Receptor (NTRK) Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
40.9kDa | |
RUO - research use only | |
NTRK2 (TRKB) | |
Human | |
His Tag | |
Frozen |