missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTAN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93298-0.02ml
This item is not returnable.
View return policy
Description
NTAN1 Polyclonal antibody specifically detects NTAN1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NTAN1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| DKFZp666E058, EC 3.5.1.-, N-terminal asparagine amidase, N-terminal asparagine amidohydrolase, PNAA, PNAD, Protein NH2-terminal asparagine amidohydrolase, Protein NH2-terminal asparagine deamidase, Protein N-terminal Asn amidase, protein N-terminal asparagine amidohydrolase, Protein NTN-amidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human NTAN1 (NP_775745.1). MPLLVEGRRVRLPQSAGDLVRAHPPLEERARLLRGQSVQQVGPQGLLYVQQRELAVTSPKDGSISILGSDDATTCHIVVLRHTGNGATCLTHCDGTDTKAEVPLIMNSIKSFSDHAQCGRLEVHLVGGFSDDRQLSQKLTHQLLSEFDRQEDDIHLVTLCVTELNDREENENHFPVIYGIAVNIKTAEIYRASFQDRGPEEQLRAARTLAGGPMISIYDAETEQLRIGPYSWTPFPHVDFWLHQDDKQILENLSTSPLAEPPHFVEHIRSTLMFLKKHPSPAHTLFSGNKALLYKKNEDGLWEKISSPGS | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 123803 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction