missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTAN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | NTAN1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655780
|
Novus Biologicals
NBP2-93298-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627850
|
Novus Biologicals
NBP2-93298-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NTAN1 Polyclonal antibody specifically detects NTAN1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NTAN1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 123803 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp666E058, EC 3.5.1.-, N-terminal asparagine amidase, N-terminal asparagine amidohydrolase, PNAA, PNAD, Protein NH2-terminal asparagine amidohydrolase, Protein NH2-terminal asparagine deamidase, Protein N-terminal Asn amidase, protein N-terminal asparagine amidohydrolase, Protein NTN-amidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human NTAN1 (NP_775745.1). MPLLVEGRRVRLPQSAGDLVRAHPPLEERARLLRGQSVQQVGPQGLLYVQQRELAVTSPKDGSISILGSDDATTCHIVVLRHTGNGATCLTHCDGTDTKAEVPLIMNSIKSFSDHAQCGRLEVHLVGGFSDDRQLSQKLTHQLLSEFDRQEDDIHLVTLCVTELNDREENENHFPVIYGIAVNIKTAEIYRASFQDRGPEEQLRAARTLAGGPMISIYDAETEQLRIGPYSWTPFPHVDFWLHQDDKQILENLSTSPLAEPPHFVEHIRSTLMFLKKHPSPAHTLFSGNKALLYKKNEDGLWEKISSPGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title