missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN2 Mouse anti-Human, Clone: 2H5A2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NSUN2 |
|---|---|
| Clone | 2H5A2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868585
|
Proteintech
66580-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858585
|
Proteintech
66580-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NSUN2 encodes a methyltransferase that catalyzes the intron-dependent formation of 5-methylcytosine at C34 of tRNA-leu(CAA). Maturation of cytoplasmic tRNAs includes splicing of introns, which are located 1 nucleotide 3-prime from the anticodon in all intron-containing tRNA genes. In tRNA-leu(CAA), the first position of the anticodon, C34, is converted to 5-methylcytosine, a modification necessary to stabilize the anticodon-codon pairing and correctly translate the mRNA. Mutations of NSUN2 result in autosomal recessive 5 mental retardation (MRT5).Specifications
| NSUN2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hTrm4, MISU, NSUN2, SAKI, TRM4 | |
| NSUN2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2H5A2 | |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q08J23 | |
| 54888 | |
| NSUN2 Fusion Protein Ag14791 | |
| Primary | |
| -20°C | |
| NSUN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title