missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN2 Mouse anti-Human, Clone: 2H5A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66580-1-IG-150UL
This item is not returnable.
View return policy
Description
NSUN2 encodes a methyltransferase that catalyzes the intron-dependent formation of 5-methylcytosine at C34 of tRNA-leu(CAA). Maturation of cytoplasmic tRNAs includes splicing of introns, which are located 1 nucleotide 3-prime from the anticodon in all intron-containing tRNA genes. In tRNA-leu(CAA), the first position of the anticodon, C34, is converted to 5-methylcytosine, a modification necessary to stabilize the anticodon-codon pairing and correctly translate the mRNA. Mutations of NSUN2 result in autosomal recessive 5 mental retardation (MRT5).Specifications
| NSUN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q08J23 | |
| NSUN2 | |
| NSUN2 Fusion Protein Ag14791 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2H5A2 | |
| Unconjugated | |
| NSUN2 | |
| hTrm4, MISU, NSUN2, SAKI, TRM4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 54888 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction