Learn More
Thermo Scientific™ NSC 23766
Brand: Thermo Scientific™ J64979.MA
Code : NEW
Additional Details : CAS Number : 733767-34-5 Weight : 0.00010kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
A cell-permeable pyrimidine compound that specifically and reversibly inhibits Rac1 GDP/GTP exchange activity.Specifications
10 mg | |
2°C to 8°C | |
733767-34-5 | |
530.96g/mol | |
Light Sensitive and Hygroscopic | |
Powder |
White | |
A cell-permeable pyrimidine compound that specifically and reversibly inhibits Rac1 GDP/GTP exchange activity | |
C24H35N7·3HCl | |
Rac1 Inhibitor | |
Signal Transduction Reagent |