missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 5806
GREENER_CHOICE

Product Code. 18074142
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074142

Brand: Tocris Bioscience™ 4166/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV4.3 channel activator

KV4.3 channel activator; mediates the transient outward K+ current (Ito). Increases IKv4.3 peak current amplitude in CHO-K1 cells expressing KV4.3 and KChIP2 (EC50 = 5.3 μM). Inhibits KV1.4-mediated currents independently of KChIP2. Also slows the decay of KV4.2 and KV4.3 currents in the presence of KChIP2.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 426834-69-7
Molecular Formula C16H8Br2F6N6O

Specifications

Quantity 50 mg
Formula Weight 574.07
Purity >99%
Molecular Weight (g/mol) 574.079
Chemical Name or Material NS 5806
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.