CAS RN 426834-69-7
CAS RN 426834-69-7
IUPAC Name:
3-[3,5-bis(trifluoromethyl)phenyl]-1-[2,4-dibromo-6-(2H-1,2,3,4-tetrazol-5-yl)phenyl]urea
Synonyms:
3-[3,5-bis(trifluoromethyl)phenyl]-1-[2,4-dibromo-6-(2H-1,2,3,4-tetrazol-5-yl)phenyl]urea
Molecular Weight (g/mol):
574.08
Molecular Formula:
C16H8Br2F6N6O
InChi Key:
UZWJWROOLOOCPQ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(=CC(NC(=O)NC2=C(Br)C=C(Br)C=C2C2=NNN=N2)=C1)C(F)(F)F