missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 5806

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
KV4.3 channel activator; mediates the transient outward K+ current (Ito). Increases IKv4.3 peak current amplitude in CHO-K1 cells expressing KV4.3 and KChIP2 (EC50 = 5.3 μM). Inhibits KV1.4-mediated currents independently of KChIP2. Also slows the decay of KV4.2 and KV4.3 currents in the presence of KChIP2.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 574.07 |
| Purity | >99% |
| Molecular Weight (g/mol) | 574.079 |
| Chemical Name or Material | NS 5806 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction