missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRXN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55051-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to NRXN1 alpha.
Neuronal cell surface protein that may be involved in cell recognition and cell adhesion by forming intracellular junctions through binding to neuroligins. May play a role in formation or maintece of synaptic junctions. May mediate intracellular signaling. May play a role in angiogenesis.Specifications
| NRXN1 | |
| Polyclonal | |
| Unconjugated | |
| NRXN1 | |
| Hs.22998, KIAA0578, neurexin 1, Neurexin 1 alpha, Neurexin I alpha, NRXN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9378 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ULB1 | |
| NRXN1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction