missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRXN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NRXN1 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16896485
|
Proteintech
55051-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16886485
|
Proteintech
55051-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to NRXN1 alpha.
Neuronal cell surface protein that may be involved in cell recognition and cell adhesion by forming intracellular junctions through binding to neuroligins. May play a role in formation or maintece of synaptic junctions. May mediate intracellular signaling. May play a role in angiogenesis.Specifications
| NRXN1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9ULB1 | |
| 9378 | |
| Peptide | |
| Primary | |
| -20°C | |
| NRXN1 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Hs.22998, KIAA0578, neurexin 1, Neurexin 1 alpha, Neurexin I alpha, NRXN1 | |
| NRXN1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title