missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRN1L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94688-0.1ml
This item is not returnable.
View return policy
Description
NRN1L Polyclonal antibody specifically detects NRN1L in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NRN1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC118990, MGC118993, MRCC2446, neuritin 1-like, neuritin-like protein, UNQ2446 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 39-139 of human NRN1L (Q496H8). PNRCDTIYQGFAECLIRLGDSMGRGGELETICRSWNDFHACASQVLSGCPEEAAAVWESLQQEARQAPRPNNLHTLCGAPVHVRERGTGSETNQETLRATA | |
| 0.1 mL | |
| Cell Biology | |
| 123904 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction