missing translation for 'onlineSavingsMsg'
Learn More

NRF2/NFE2L2 Mouse anti-Human, Clone: 1E9E3, Proteintech

Mouse Monoclonal Antibody

£122.57 - £298.00

Specifications

Antigen NRF2/NFE2L2
Clone 1E9E3
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808445
View Documents
Proteintech
66504-1-IG-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16898435
View Documents
Proteintech
66504-1-IG-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Nrf2 (nuclear factor erythroid 2-related factor 2) is a transcription activator which plays a key role in protecting the cell from oxidative stress and inflammation. It is primarily localized in the cytoplasm under normal conditions, but under conditions of oxidative stress, Nrf2 migrates to the nucleus to upregulate expression of genes with ARE (antioxidant response elements) in the promoter region. The upregulated genes include those involved in cellular protection, drug transport, detoxification, and anti-oxidant defense. The protein is ubiquitously expressed in various tissues, with the highest levels found in kidney, muscle, lung, heart, liver, and brain.
Specifications

Specifications

NRF2/NFE2L2
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HEBP1, NF E2 related factor 2, NFE2 related factor 2, NFE2L2, NRF2
NFE2L2
IgG2b
Protein A
Antibody
1E9E3
Western Blot
Unconjugated
Mouse
Human
Q16236
4780
NRF2, NFE2L2 Fusion Protein Ag9469
Primary
-20°C
NFE2L2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.