missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRF2/NFE2L2 Mouse anti-Human, Clone: 1E9E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66504-1-IG-150UL
This item is not returnable.
View return policy
Description
Nrf2 (nuclear factor erythroid 2-related factor 2) is a transcription activator which plays a key role in protecting the cell from oxidative stress and inflammation. It is primarily localized in the cytoplasm under normal conditions, but under conditions of oxidative stress, Nrf2 migrates to the nucleus to upregulate expression of genes with ARE (antioxidant response elements) in the promoter region. The upregulated genes include those involved in cellular protection, drug transport, detoxification, and anti-oxidant defense. The protein is ubiquitously expressed in various tissues, with the highest levels found in kidney, muscle, lung, heart, liver, and brain.Specifications
| NRF2/NFE2L2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16236 | |
| NFE2L2 | |
| NRF2, NFE2L2 Fusion Protein Ag9469 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 1E9E3 | |
| Unconjugated | |
| NFE2L2 | |
| HEBP1, NF E2 related factor 2, NFE2 related factor 2, NFE2L2, NRF2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4780 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction